4-Fluoro-2-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

F1052-1G F1052-5G
TCF1052-1GEA 96.38 CAD
TCF1052-1G TCF1052-5G
4-Fluoro-2-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
4-Fluoro-2-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BF₄O₂
MDL Number: MFCD08062379
CAS Number: 182344-16-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR