2-Fluoro-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

F1006-1G F1006-5G
TCF1006-1GEA 47.87 CAD
TCF1006-1G TCF1006-5G
2-Fluoro-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
2-Fluoro-3-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BF₄O₂
MDL Number: MFCD07784336
CAS Number: 157834-21-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 95.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR