3,5-Dibromo-p-toluidine ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
3,5-Dibromo-4-methylaniline
D4730-1G
D4730-5G
TCD4730-1GEA
281.24
CAD
TCD4730-1G
TCD4730-5G
3,5-Dibromo-p-toluidine ≥97.0% (by GC, titration analysis)
3,5-Dibromo-p-toluidine
Formula:
C₇H₇Br₂N MW: 264.96 g/mol |
MDL Number:
MFCD00151806 CAS Number: 13194-73-5 |
Specification Test Results
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 88.0 to 92.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...