4-(Pentyloxy)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

P2358-1G P2358-5G
TCP2358-1GEA 176.96 CAD
TCP2358-1G TCP2358-5G
4-(Pentyloxy)phenylboronic acid (contains varying amounts of Anhydride)
4-(Pentyloxy)phenylboronic acid
Formula: C₁₁H₁₇BO₃
Melting Pt: 80°C
MDL Number: MFCD04039035
CAS Number: 146449-90-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR