4-Hexylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

H1489-1G H1489-5G
TCH1489-1GEA 116.92 CAD
TCH1489-1G TCH1489-5G
4-Hexylphenylboronic acid (contains varying amounts of Anhydride)
4-Hexylphenylboronic acid
Formula: C₁₂H₁₉BO₂
MDL Number: MFCD04039029
CAS Number: 105365-50-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR