5'-Bromo-2'-hydroxy-3'-nitroacetophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1-(5-Bromo-2-hydroxy-3-nitrophenyl)ethanone

B4556-25G B4556-5G
TCB4556-25GEA 417.12 CAD
TCB4556-25G TCB4556-5G
5'-Bromo-2'-hydroxy-3'-nitroacetophenone ≥98.0% (by GC, titration analysis)
5'-Bromo-2'-hydroxy-3'-nitroacetophenone
Formula: C₈H₆BrNO₄
MDL Number: MFCD01631134
CAS Number: 70978-54-0

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 129.0 to 133.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR