3-Chloro-N,N-dimethylaniline ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
C3192-25G
C3192-5G
TCC3192-25GEA
508.76
CAD
TCC3192-25G
TCC3192-5G
3-Chloro-N,N-dimethylaniline ≥97.0% (by GC, titration analysis)
3-Chloro-N,N-dimethylaniline
Formula:
C₈H₁₀ClN Boiling Pt: 234°C |
MDL Number:
MFCD00018456 CAS Number: 6848-13-1 EINECS: 229-935-4 |
Specification Test Results
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...