1,1,1-Trifluoroacetone cyanohydrin ≥96.0% (by GC)

Supplier: TCI America
Danger

H1508-1G H1508-5G
TCH1508-1GEA 112.18 CAD
TCH1508-1G TCH1508-5G
1,1,1-Trifluoroacetone cyanohydrin ≥96.0% (by GC)
1,1,1-Trifluoroacetone cyanohydrin
Formula: C₄H₄F₃NO
Boiling Pt: 139°C
Flash Pt: 60°C
MDL Number: MFCD00040885
CAS Number: 335-08-0
UN: 3275
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR