1,3-Dimethyl-1H-pyrazole-5-carboxylic acid ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
D5468-25G
D5468-5G
76220-000EA
436.08
CAD
76220-000
76220-002
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid ≥98.0% (by GC)
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid
Formula:
C₆H₈N₂O₂ |
MDL Number:
MFCD00159636 CAS Number: 5744-56-9 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Melting point | 208.0 to 212.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...