2-Amino-6-chloro-3-formylchromone ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Warning
|
A2743-1G
A2743-5G
TCA2743-1GEA
84.37
CAD
TCA2743-1G
TCA2743-5G
2-Amino-6-chloro-3-formylchromone ≥98.0% (by HPLC, total nitrogen)
2-Amino-6-chloro-3-formylchromone
Formula:
C₁₀H₆ClNO₃ Melting Pt: 290°C |
MDL Number:
MFCD00139140 CAS Number: 68301-77-9 |
Specification Test Results
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...