Tetrathiafulvalene ≥99.0% (by GC), purified by sublimation

Supplier: TCI America

T3380-1G T3380-200MG
TCT3380-1GEA 298.87 CAD
TCT3380-1G TCT3380-200MG
Tetrathiafulvalene ≥99.0% (by GC), purified by sublimation
Tetrathiafulvalene
Formula: C₆H₄S₄
MDL Number: MFCD00005492
CAS Number: 31366-25-3
EINECS: 250-593-7
Merck Index: 14,09242

Order Now

Specification Test Results

Appearance Orange to Brown powder to crystal
Purity (GC) min. 99.0 %
Melting point 118.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR