4-Chloro-3-methylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
C3071-1G
C3071-5G
TCC3071-1GEA
83.74
CAD
TCC3071-1G
TCC3071-5G
4-Chloro-3-methylphenylboronic acid (contains varying amounts of Anhydride)
4-Chloro-3-methylphenylboronic acid
Formula:
C₇H₈BClO₂ Melting Pt: 258°C |
MDL Number:
MFCD02683104 CAS Number: 161950-10-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 112.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...