2-Chloro-5-methylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
C3211-1G
C3211-5G
TCC3211-1GEA
189.6
CAD
TCC3211-1G
TCC3211-5G
2-Chloro-5-methylphenylboronic acid (contains varying amounts of Anhydride)
2-Chloro-5-methylphenylboronic acid
Formula:
C₇H₈BClO₂ |
MDL Number:
MFCD06659822 CAS Number: 193353-35-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 111.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...