4',5'-Dichloro-2'-nitroacetanilide ≥97.0% (by GC)

Supplier: TCI America
Warning

D4907-25G D4907-5G
TCD4907-25GEA 587.76 CAD
TCD4907-25G TCD4907-5G
4',5'-Dichloro-2'-nitroacetanilide ≥97.0% (by GC)
4',5'-Dichloro-2'-nitroacetanilide
Formula: C₈H₆Cl₂N₂O₃
MDL Number: MFCD00024301
CAS Number: 5462-30-6

Order Now

Specification Test Results

Purity (GC) min. 97.0 %
Melting point 124.0 to 128.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR