2-Methoxy-4-nitrobenzonitrile ≥97.0% (by GC)

Supplier: TCI America
Danger

M2585-1G M2585-5G
TCM2585-1GEA 53.72 CAD
TCM2585-1G TCM2585-5G
2-Methoxy-4-nitrobenzonitrile ≥97.0% (by GC)
2-Methoxy-4-nitrobenzonitrile
Formula: C₈H₆N₂O₃
MW: 178.15 g/mol
MDL Number: MFCD02093781
CAS Number: 101084-96-2
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 177.0 to 181.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR