Ethyl-2-aminonicotinate ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

E1146-1G E1146-5G
TCE1146-1GEA 52.14 CAD
TCE1146-1G TCE1146-5G
Ethyl-2-aminonicotinate ≥98.0% (by GC, titration analysis)
Ethyl 2-aminonicotinate
Formula: C₈H₁₀N₂O₂
Boiling Pt: 133°C
MDL Number: MFCD01569452
CAS Number: 13362-26-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 93.0 to 97.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR