3-Bromo-5-nitroanisole ≥98.0% (by GC)

Supplier: TCI America
Warning

B4973-1G B4973-5G
TCB4973-1GEA 126.4 CAD
TCB4973-1G TCB4973-5G
3-Bromo-5-nitroanisole ≥98.0% (by GC)
3-Bromo-5-nitroanisole
Formula: C₇H₆BrNO₃
MDL Number: MFCD04112752
CAS Number: 16618-67-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 86.0 to 90.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR