Methyl (3-bromophenyl)acetate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Bromophenylacetic acid methyll ester, Methyl 3-Bromophenylacetate, Methyl 2-(3-bromophenyl)acetate

M2804-25G M2804-5G
TCM2804-25GEA 364.98 CAD
TCM2804-25G TCM2804-5G
Methyl (3-bromophenyl)acetate ≥98.0% (by GC)
Methyl (3-bromophenyl)acetate
Formula: C₉H₉BrO₂
MW: 229.0700 g/mol
MDL Number: MFCD06858764
CAS Number: 150529-73-0

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR