Dimethyl-3-nitrophthalate ≥99.0% (by GC)

Supplier: TCI America

Synonyms: Dimethyl 3-nitrophthalate

D5310-25G D5310-5G
TCD5310-25GEA 739.44 CAD
TCD5310-25G TCD5310-5G
Dimethyl-3-nitrophthalate ≥99.0% (by GC)
Dimethyl 3-nitrophthalate
Formula: C₁₀H₉NO₆
MW: 239.19 g/mol
MDL Number: MFCD00017184
CAS Number: 13365-26-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 66.0 to 70.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR