2,3-Difluoroisonicotinic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
D5131-1G
D5131-5G
76219-172EA
44.24
CAD
76219-172
76219-174
2,3-Difluoroisonicotinic acid ≥98.0% (by GC, titration analysis)
2,3-Difluoroisonicotinic acid
Formula:
C₆H₃F₂NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD09031174 CAS Number: 851386-31-7 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...