4-Methyl-3-(trifluoromethyl)benzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M2708-1G M2708-5G
TCM2708-1GEA 64.78 CAD
TCM2708-1G TCM2708-5G
4-Methyl-3-(trifluoromethyl)benzoic acid ≥98.0% (by GC, titration analysis)
4-Methyl-3-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₂
MDL Number: MFCD01631592
CAS Number: 261952-01-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 175.0 to 179.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR