2-Mercaptopyridine-N-oxide sodium salt, anhydrous ≥98.0% (by HPLC)

Supplier: TCI America
Warning

M2841-25G M2841-5G
TCM2841-25GEA 478.74 CAD
TCM2841-25G TCM2841-5G
2-Mercaptopyridine-N-oxide sodium salt, anhydrous ≥98.0% (by HPLC)
2-Mercaptopyridine N-oxide sodium salt
Formula: C₅H₄NNaOS
MDL Number: MFCD01941547
CAS Number: 3811-73-2
EINECS: 223-296-5
Merck Index: 14,07994

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Iodometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR