3,5-Dimethyl-4-propoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

D5080-1G D5080-5G
TCD5080-1GEA 45.82 CAD
TCD5080-1G TCD5080-5G
3,5-Dimethyl-4-propoxyphenylboronic acid (contains varying amounts of Anhydride)
3,5-Dimethyl-4-propoxyphenylboronic acid
Formula: C₁₁H₁₇BO₃
MDL Number: MFCD05865178
CAS Number: 357611-51-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR