3,5-Dihydroxy-4-methylbenzoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
D4712-1G
D4712-5G
TCD4712-1GEA
74.26
CAD
TCD4712-1G
TCD4712-5G
3,5-Dihydroxy-4-methylbenzoic acid ≥98.0% (by titrimetric analysis)
3,5-Dihydroxy-4-methylbenzoic acid
Formula:
C₈H₈O₄ |
MDL Number:
MFCD00002514 CAS Number: 28026-96-2 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...