2-Chloro-4-methylphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

C3049-1G C3049-5G
TCC3049-1GEA 42.66 CAD
TCC3049-1G TCC3049-5G
2-Chloro-4-methylphenylboronic acid (contains varying amounts of Anhydride)
2-Chloro-4-methylphenylboronic acid
Formula: C₇H₈BClO₂
Melting Pt: 202°C
MDL Number: MFCD03411936
CAS Number: 145349-62-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR