Methyl-4-ethynylbenzoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 4-ethynylbenzoate

M2768-1G M2768-5G
TCM2768-1GEA 203.82 CAD
TCM2768-1G TCM2768-5G
Methyl-4-ethynylbenzoate ≥98.0% (by GC)
Methyl 4-ethynylbenzoate
Formula: C₁₀H₈O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00168820
CAS Number: 3034-86-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 97.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR