2-Amino-4-(2,4-difluorophenyl)thiazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A3029-1G A3029-5G
TCA3029-1GEA 127.98 CAD
TCA3029-1G TCA3029-5G
2-Amino-4-(2,4-difluorophenyl)thiazole ≥98.0% (by GC, titration analysis)
2-Amino-4-(2,4-difluorophenyl)thiazole
Formula: C₉H₆F₂N₂S
MDL Number: MFCD00472891
CAS Number: 105512-80-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 116.0 to 120.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR