2-Amino-4-(4-chlorophenyl)thiazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

A2841-1G A2841-5G
TCA2841-1GEA 97.96 CAD
TCA2841-1G TCA2841-5G
2-Amino-4-(4-chlorophenyl)thiazole ≥98.0% (by GC, titration analysis)
2-Amino-4-(4-chlorophenyl)thiazole
Formula: C₉H₇ClN₂S
MDL Number: MFCD00047058
CAS Number: 2103-99-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 167.0 to 171.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR