Pentafluorophenyl 4-nitrobenzenesulfonate ≥98.0% (by GC)

Supplier: TCI America
Warning

P2231-1G P2231-5G
TCP2231-1GEA 95.91 CAD
TCP2231-1G TCP2231-5G
Pentafluorophenyl 4-nitrobenzenesulfonate ≥98.0% (by GC)
Pentafluorophenyl 4-nitrobenzenesulphonate
Formula: C₁₂H₄F₅NO₅S
MDL Number: MFCD05975120
CAS Number: 244633-31-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 106.0 to 110.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR