3-Amino-3-(4-methoxyphenyl)propionic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-Amino-3-(4-methoxyphenyl)propionic Acid, 3-Amino-3-(4-methoxyphenyl)propanoic acid

A2532-1G A2532-5G
TCA2532-1GEA 214.88 CAD
TCA2532-1G TCA2532-5G
3-Amino-3-(4-methoxyphenyl)propionic acid ≥98.0% (by HPLC, titration analysis)
3-Amino-3-(4-methoxyphenyl)propionic acid
Formula: C₁₀H₁₃NO₃
Melting Pt: 232°C
MDL Number: MFCD00187209
CAS Number: 5678-45-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR