2'-Cyanoacetanilide ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Cyanoacetanilide, 2-Cyano-N-phenylacetamide

C2894-1G C2894-5G
TCC2894-1GEA 135.88 CAD
TCC2894-1G TCC2894-5G
2'-Cyanoacetanilide ≥98.0% (by GC)
2'-Cyanoacetanilide
Formula: C₉H₈N₂O
MW: 160.1726 g/mol
MDL Number: MFCD00019840
CAS Number: 621-03-4
EINECS: 210-666-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 198.0 to 202.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR