3,5-Dibromo-1H-1,2,4-triazole ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
3,5-Dibromo-1,2,4-triazole
D5030-25G
D5030-5G
TCD5030-25GEA
521.4
CAD
TCD5030-25G
TCD5030-5G
3,5-Dibromo-1H-1,2,4-triazole ≥97.0% (by GC)
3,5-Dibromo-1H-1,2,4-triazole
Formula:
C₂HBr₂N₃ MW: 226.8600 g/mol |
MDL Number:
MFCD00487260 CAS Number: 7411-23-6 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...