2,4-Dimethyl-5-thiazolecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2,4-Dimethylthiazole-5-carboxylic acid

D4813-1G D4813-5G
TCD4813-1GEA 164.32 CAD
TCD4813-1G TCD4813-5G
2,4-Dimethyl-5-thiazolecarboxylic acid ≥98.0% (by GC, titration analysis)
2,4-Dimethyl-5-thiazolecarboxylic acid
Formula: C₆H₇NO₂S
MDL Number: MFCD00052943
CAS Number: 53137-27-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR