3-Bromo-6-chlorochromone ≥98.0% (by GC)

Supplier: TCI America
Warning

B4967-1G B4967-5G
TCB4967-1GEA 137.46 CAD
TCB4967-1G TCB4967-5G
3-Bromo-6-chlorochromone ≥98.0% (by GC)
3-Bromo-6-chlorochromone
Formula: C₉H₄BrClO₂
MDL Number: MFCD02093123
CAS Number: 73220-38-9

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 129.0 to 133.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR