2-Amino-4-(1-naphthyl)thiazole ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
4-(Naphthalen-1-yl)thiazol-2-amine
A2888-1G
A2888-5G
TCA2888-1GEA
112.34
CAD
TCA2888-1G
TCA2888-5G
2-Amino-4-(1-naphthyl)thiazole ≥97.0% (by GC, titration analysis)
2-Amino-4-(1-naphthyl)thiazole
Formula:
C₁₃H₁₀N₂S MW: 226.30 g/mol |
MDL Number:
MFCD00236016 CAS Number: 56503-96-9 |
Specification Test Results
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 156.0 to 160.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...