2-Methyl-4-phenylthiazol ≥98.0% (by GC)

Supplier: TCI America

M2892-1G M2892-5G
TCM2892-1GEA 71.1 CAD
TCM2892-1G TCM2892-5G
2-Methyl-4-phenylthiazol ≥98.0% (by GC)
2-Methyl-4-phenylthiazol
Formula: C₁₀H₉NS
Boiling Pt: 284°C
MDL Number: MFCD00489998
CAS Number: 1826-16-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 68.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR