Cyclohexyldiphenylphosphine oxide ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
C3310-1G
C3310-5G
TCC3310-1GEA
39.5
CAD
TCC3310-1G
TCC3310-5G
Cyclohexyldiphenylphosphine oxide ≥98.0% (by GC)
Cyclohexyldiphenylphosphine oxide
Formula:
C₁₈H₂₁OP MW: 284.34 g/mol |
MDL Number:
MFCD00014301 CAS Number: 13689-20-8 EINECS: 237-204-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 167.0 to 171.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...