(±)-1-Benzyl-3-hydroxypiperidine ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B4868-25G B4868-5G
TCB4868-25GEA 423.44 CAD
TCB4868-25G TCB4868-5G
(±)-1-Benzyl-3-hydroxypiperidine ≥97.0% (by GC, titration analysis)
(±)-1-Benzyl-3-hydroxypiperidine
Formula: C₁₂H₁₇NO
Boiling Pt: 153°C
MDL Number: MFCD00023734
CAS Number: 14813-01-5
EINECS: 238-881-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR