3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3-(4-Fluorophenyl)-5-methylisoxazol-4-carboxylic acid

F1023-1G F1023-5G
TCF1023-1GEA 80.58 CAD
TCF1023-1G TCF1023-5G
3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid ≥98.0% (by GC, titration analysis)
3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid
Formula: C₁₁H₈FNO₃
MW: 221.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03407356
CAS Number: 1736-21-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 200.0 to 204.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR