2-Ethoxy-5-pyridineboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Ethoxy-5-pyridylboronic acid, 6-Ethoxy-3-pyridylboronic acid, 2-Ethoxypyridine-5-boronic acid, (6-Ethoxypyridin-3-yl)boronic acid, 6-Ethoxypyridine-3-boronic acid
E1115-1G
E1115-5G
TCE1115-1GEA
47.4
CAD
TCE1115-1G
TCE1115-5G
2-Ethoxy-5-pyridineboronic acid (contains varying amounts of Anhydride)
2-Ethoxy-5-pyridineboronic acid
Formula:
C₇H₁₀BNO₃ MW: 166.97 g/mol Melting Pt: 134°C |
MDL Number:
MFCD06201032 CAS Number: 612845-44-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 113.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...