Cyclopentanecarbaldehyde 90,0 GC_ASSAY_METHOD Stabilised

Supplier: TCI America
Warning

Synonyms: Cyclopentanecarboxaldehyde

Stabilization: Hydrochinon

C3019-1ML C3019-5ML
TCC3019-1MLEA 91.64 CAD
TCC3019-1ML TCC3019-5ML
Cyclopentanecarbaldehyde 90,0 GC_ASSAY_METHOD Stabilised
Cyclopentanecarbaldehyde
Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 40°C
Flash Pt: 28°C
Storage Temperature: Refrigerator
MDL Number: MFCD00060798
CAS Number: 872-53-7
EINECS: 212-829-7
UN: 1989
ADR: 3 III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR