4-(4-Bromophenoxy)benzaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

B4900-1G B4900-5G
TCB4900-1GEA 146.94 CAD
TCB4900-1G TCB4900-5G
4-(4-Bromophenoxy)benzaldehyde ≥98.0% (by GC)
4-(4-Bromophenoxy)benzaldehyde
Formula: C₁₃H₉BrO₂
Boiling Pt: 189°C
MDL Number: MFCD08276811
CAS Number: 69240-56-8

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR