3,5-Dimethylisoxazole-4-carbonyl chloride ≥96.0% (by GC, titration analysis)

Supplier: TCI America
Danger

D4706-1G D4706-5G
TCD4706-1GEA 107.44 CAD
TCD4706-1G TCD4706-5G
3,5-Dimethylisoxazole-4-carbonyl chloride ≥96.0% (by GC, titration analysis)
3,5-Dimethylisoxazole-4-carbonyl chloride
Formula: C₆H₆ClNO₂
Boiling Pt: 90°C
MDL Number: MFCD00052555
CAS Number: 31301-45-8
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %
Purity (Argentmetric Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR