6-Bromochromone-2-carboxylic acid ≥95.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B4802-1G B4802-5G
TCB4802-1GEA 61.62 CAD
TCB4802-1G TCB4802-5G
6-Bromochromone-2-carboxylic acid ≥95.0% (by GC, titration analysis)
6-Bromochromone-2-carboxylic acid
Formula: C₁₀H₅BrO₄
Melting Pt: 276°C
MDL Number: MFCD01242619
CAS Number: 51484-06-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR