1-Phenylcyclohexanecarboxylic acid ≥95.0% (by GC, titration analysis)

Supplier: TCI America
Warning

P2180-1G P2180-5G
TCP2180-1GEA 53.72 CAD
TCP2180-1G TCP2180-5G
1-Phenylcyclohexanecarboxylic acid ≥95.0% (by GC, titration analysis)
1-Phenylcyclohexanecarboxylic acid
Formula: C₁₃H₁₆O₂
MDL Number: MFCD00037152
CAS Number: 1135-67-7
EINECS: 214-495-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 121.0 to 125.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR