1-Ethyl-1-methylpyrrolidinium bromide ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

E1050-25G E1050-5G
TCE1050-25GEA 101.12 CAD
TCE1050-25G TCE1050-5G
1-Ethyl-1-methylpyrrolidinium bromide ≥97.0% (by titrimetric analysis)
1-Ethyl-1-methylpyrrolidinium bromide
Formula: C₇H₁₆BrN
MDL Number: MFCD03095384
CAS Number: 69227-51-6
EINECS: 418-200-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR