2-Methyl-5-nitrobenzotrifluoride ≥98.0% (by GC)
Supplier: TCI America
|
Danger
|
M2798-1G
M2798-5G
TCM2798-1GEA
33.18
CAD
TCM2798-1G
TCM2798-5G
2-Methyl-5-nitrobenzotrifluoride ≥98.0% (by GC)
2-Methyl-5-nitrobenzotrifluoride
Formula:
C₈H₆F₃NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD01631684 CAS Number: 89976-12-5 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 38.0 to 42.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...