4-Bromo-1-naphthoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B4949-1G B4949-5G
TCB4949-1GEA 61.62 CAD
TCB4949-1G TCB4949-5G
4-Bromo-1-naphthoic acid ≥98.0% (by GC, titration analysis)
4-Bromo-1-naphthoic acid
Formula: C₁₁H₇BrO₂
MDL Number: MFCD00094221
CAS Number: 16650-55-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 223.0 to 227.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR