(4-Aminobenzyl)phosphonic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: (4-Aminobenzyl)phosphonic Acid

A3072-1G A3072-200MG
76218-786EA 497.7 CAD
76218-786 76218-788
(4-Aminobenzyl)phosphonic acid ≥98.0% (by HPLC, titration analysis)
(4-Aminobenzyl)phosphonic acid
Formula: C₇H₁₀NO₃P
Melting Pt: 325°C
Storage Temperature: Freezer
MDL Number: MFCD00070204
CAS Number: 5424-27-1

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR