4-Acetamidobenzyl alcohol ≥98.0% (by GC)

Supplier: TCI America

H1605-1G H1605-5G
TCH1605-1GEA 79 CAD
TCH1605-1G TCH1605-5G
4-Acetamidobenzyl alcohol ≥98.0% (by GC)
4-Acetamidobenzyl alcohol
Formula: C₉H₁₁NO₂
MDL Number: MFCD00016868
CAS Number: 16375-88-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 125.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR